Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Racepinephrine Hydrochloride, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
329-63-5
C9H14ClNO3
219.67
ATADHKWKHYVBTJ-UHFFFAOYNA-N
hydrogen 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol chloride
[H+].[Cl-].CNCC(O)C1=CC=C(O)C(O)=C1
329-63-5
C9H14ClNO3
219.67
ATADHKWKHYVBTJ-UHFFFAOYNA-N
hydrogen 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol chloride
[H+].[Cl-].CNCC(O)C1=CC=C(O)C(O)=C1

E1418, 329-63-5, C9H13NO3•HCl

Spectrum™ Racepinephrine Hydrochloride, USP is a bronchodilator used to relieve occasional symptoms of asthma, such as wheezing, chest tightness, and feeling short of breath. All Spectrum Chemical USP products, including all USP hydrochloride products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Water 0.005
Ignition Residue 0.005
Specific Rotation -1° to +1°
Quantity 5 g
CAS Max % 1
Percent Purity 97 to 102%
Grade USP