Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Racked Filter Pipette Tips
SDPSCI_ED

Catalog No. S139630 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
10 μL
100 μL
200 μL
300 μL
1000 μL
1250 μL
Catalog No. Volume (Metric)
S139630 300 μL
S111108 10 μL
S111109 100 μL
S111110 200 μL
S111111 1000 μL
S111112 1250 μL
6 options
This item is not returnable. View return policy

Catalog No. S139630

Supplier: United Scientific™ UNUFT0300UL

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

These filter pipette tips are sterile, racked, and laboratory approved.

  • Protect against cross-contamination using these racked, sterile filter pipette tips
  • These universal tips are manufactured from polypropylene and the filters are manufactured from polyethylene with a hydrophobic design to prevent liquids and aerosols from entering the body of the micropipette
  • These tips are non-cytotoxic, non-hemolytic, and have 99.9% Bacterial Aerosol Filtration Efficiency (BFE)
  • DNase, RNase and Pyrogen-free

Specifications

Volume (Metric) 300 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model UNUFT0300UL
For Use With (Application) Pipetting
Format Rack, 10 x 96
Tip Style Standard
Packaging Racked
Product Type Filter Pipette Tips
For Use With (Equipment) Universal Fit
Material Polypropylene
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.