Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Racked Low Retention Filter Pipette Tips
SDPSCI_ED

These low retention filter pipette tips are sterile, racked, and laboratory approved.

$126.00 - $132.00

Specifications

Filtered Filtered
Sterility Sterile
Color Clear
For Use With (Application) Pipetting
Format Rack, 10 x 96
View More Specs

Products 6
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
S111113
SDPSCI_ED
View Documents
United Scientific™
UNUFT010ULLR
10 μL
Case of 960 for $126.00
Add to Cart
 
S111114
SDPSCI_ED
View Documents
United Scientific™
UNUFT0100ULLR
100 μL
Case of 960 for $127.00
Add to Cart
 
S111115
SDPSCI_ED
View Documents
United Scientific™
UNUFT0200ULLR
200 μL
Case of 960 for $128.00
Add to Cart
 
S139631
SDPSCI_ED
View Documents
United Scientific™
UNUFT0300ULLR
300 μL
Pack of 960 for $130.00
Add to Cart
 
S111116
SDPSCI_ED
View Documents
United Scientific™
UNUFT01000ULLR
1000 μL
Case of 960 for $130.00
Add to Cart
 
S111117
SDPSCI_ED
View Documents
United Scientific™
UNUFT01250ULLR
1250 μL
Case of 960 for $132.00
Add to Cart
 
Description

Description

  • Protect against cross-contamination using these racked, sterile low retention filter pipette tips
  • Retention level is 10 times lesser with viscous samples than standard pipette tips
  • These universal tips are manufactured from polypropylene and the filters are manufactured from polyethylene with a hydrophobic design to prevent liquids and aerosols from entering the body of the micropipette
  • These tips are non-cytotoxic, non-hemolytic, and have 99.9% Bacterial Aerosol Filtration Efficiency (BFE)
  • DNase, RNase and Pyrogen-free, packaged in racks of 96 with 10 racks in each
Specifications

Specifications

Filtered
Clear
Rack, 10 x 96
Racked
Polypropylene
Sterile
Pipetting
Standard Length
Pipettors
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit