Learn More
Thermo Scientific™ Racks for Forma™ 88000 and TSU Series Freezers Available on GSA/VA Contract for Federal Government customers only.
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for Forma™ 88000 and TSU Series Freezers.
$250.00 - $390.50
Products 7
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
1950520 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
1950520
Adjustable Side Access Rack 2 in. boxes Each for $280.00
Add to cart
1950521
View Documents
Thermo Scientific™
1950521
Adjustable Side Access Rack 3 in. boxes Each for $280.00
Add to cart
19-505-23 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
1950523
Side Access Microplate Rack Standard Plates Each for $338.50
Add to cart
1950592 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
1950592
Side Access Microplate Rack Microplates Each for $250.00
Add to cart
920090 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
920090
Sliding drawer rack 2 in. boxes Each for $261.50
Add to cart
920091 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
920091
Sliding drawer rack 3 in. boxes Each for $261.50
Add to cart
19-505-25 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
1950525
Side Access Microplate/Deepwell Rack Deepwell microplates Each for $390.50
Add to cart
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.