Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers

Maximize space using Thermo Scientific™ Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers.

Manufacturer:  Thermo Scientific™ 398333

 View more versions of this product

Catalog No. 398333


Add to cart

Description

Description

Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ ;Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.

  • Warranty varies by country, please contact us for details
Specifications

Specifications

13.5 cm
66
Silver
55.9 cm
17.3, 23 and 28.8 cu. ft. capacity upright freezers
Microplates
Side Access Rack (Microplate)
Rectangle
23.9 cm
22 in.
Side Access Microplate Rack
9.4 in.
5.3 in.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit