Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51, Mouse anti-Human, DyLight 755, Clone: RAD51/2701, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279895IR

Catalog No. NB012042


Add to cart

Description

Description

Rad51 Monoclonal specifically detects Rad51 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rad51
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
RAD51
Recombinant fragment of human Rad51 protein (around aa 1-134) (exact sequence is proprietary)
0.1 mL
Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific
Human
IgG2b κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
RAD51/2701
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit