Learn More
Invitrogen™ RAD51AP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113706
Description
Immunogen sequence: EDDVGGVQGK RKAASKAAAQ QRKILLEGSD GDSANDTEPD FAPGEDSEDD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 93%; Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 92%.
May participate in a common DNA damage response pathway associated with the activation of homologous recombination and double-strand break repair. Functionally cooperates with PALB2 in promoting of D-loop formation by RAD51. Binds to single and double stranded DNA, and is capable of aggregating DNA. Also binds RNA.
Specifications
RAD51AP1 | |
Polyclonal | |
Unconjugated | |
Rad51ap1 | |
2510006L10Rik; PIR51; RAB22; RAD51 associated protein 1; rad51ap1; RAD51-associated protein 1; RAD51-interacting protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
10635 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96B01 | |
Rad51ap1 | |
Synthetic peptide directed towards the middle region of human RAD51AP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.