Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51C Mouse anti-Human, Mouse, Primate, Yeast, PE-Cyanine7, Clone: 2H11/6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB100177PECY7

Catalog No. N10177PECY7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Rad51C Monoclonal antibody specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for Flow Cytometry.
Specifications

Specifications

Rad51C
2H11/6
Flow Cytometry
Conjugated, Purified
RAD51C
His-tagged human Rad51C, over-expressed in E. coli. [UniProt No. O43502]
Protein G purified
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
Primary
5889.0
Human, Mouse, Primate, Yeast
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit