Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD51C Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518106
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
ZFPM1 is a transcription regulator that plays an essential role in erythroid and megakaryocytic cell differentiation. ZFPM1 is an essential cofactor that acts via the formation of a heterodimer with transcription factors of the GATA family GATA1, GATA2 and GATA3. Such heterodimer can both activate or repress transcriptional activity, depending on the cell and promoter context. The heterodimer formed with GATA proteins is essential to activate expression of genes such as NFE2, ITGA2B, alpha- and beta-globin, while it represses expression of KLF1. ZFPM1 may be involved in regulation of some genes in gonads. ZFPM1 may also be involved in cardiac development, in a non-redundant way with ZFPM2/FOG2.
Specifications
RAD51C | |
Polyclonal | |
Unconjugated | |
RAD51C | |
BROVCA3; DNA repair protein RAD51 homolog 3; FANCO; MGC104277; R51H3; RAD51 homolog C; RAD51 paralog C; RAD51C; RAD51L2; RAD51-like protein 2; RGD1563765; yeast RAD51 homolog 3 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5889 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43502 | |
RAD51C | |
Synthetic peptide sequence (RGKTFRFEMQRDL) corresponding to the N-terminus amino acids of RAD51C (aa 2-15). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction