Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD51L1 Mouse anti-Human, Clone: OTI7H4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503550

Catalog No. 50-167-4414


Add to cart

Description

Description

The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are evolutionarily conserved proteins essential for DNA repair by homologous recombination. This protein has been shown to form a stable heterodimer with the family member RAD51C, which further interacts with the other family members, such as RAD51, XRCC2, and XRCC3. Overexpression of this gene was found to cause cell cycle G1 delay and cell apoptosis, which suggested a role of this protein in sensing DNA damage. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications

Specifications

RAD51L1
Monoclonal
0.27 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O15315
R51H2, RAD51L1, REC2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5890
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI7H4
Unconjugated
RAD51B
Liquid
RAD51B
Human recombit protein fragment corresponding to amino acids 119-346 of human RAD51L1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit