Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD54L Mouse anti-Human, DyLight 488, Clone: 38.4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141218G

Catalog No. NBP141218G


Add to cart

Description

Description

RAD54L Monoclonal antibody specifically detects RAD54L in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

RAD54L
38.4
Western Blot, ELISA
Q92698
EC 3.6.1, hHR54EC 3.6.4.-, HR54, hRAD54HRAD54, RAD54 (S.cerevisiae)-like, RAD54 homolog, RAD54ADNA repair and recombination protein RAD54-like, RAD54-like (S. cerevisiae)
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
This antibody will recognize human Rad 54.
ELISA, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
RAD54L
Synthetic peptide - Combination of N & C peptide - Corresponding to a range between amino acids 1-300 and 450-741.
Immunogen affinity purified
DNA Repair, Homologous Recombination
Primary
8438
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit