Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD9B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549349

Catalog No. PA549349

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RAD9B
Polyclonal
Unconjugated
RAD9B
Liquid
RAD9B
KLH conjugated synthetic peptide between 394-428 amino acids from the C-terminal region of human RAD9B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide
Q6WBX8
Cell cycle checkpoint control protein RAD9B, DNA repair exonuclease rad9 homolog B, hRAD9B, RAD9B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
144715
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit