Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RadEye™ G Series Personal Dose Rate Meters
GSA_VA

Detect and measure even the smallest change in gamma and dose rate measurements with intrinsically safe Thermo Scientific™ RadEye™ G Series of personal radiation detectors.

Manufacturer:  Thermo Scientific™ 425067460

 View more versions of this product

Catalog No. 19-155-205


Add to cart

Description

Description

Continuously monitor your radiation exposure in any nuclear or radiological emergency. The Thermo Scientific™ RadEye™ G Series of personal radiation detectors provide first responders with quick, reliable gamma and dose rate measurements. These light-weight, handheld Geiger counters detect and measure very low gamma energies; even the smallest change in radiation rates are displayed immediately, while coincidentally occurring fluctuations are suppressed. For hazmat teams investigating potentially explosive atmospheres, intrinsically safe RadEye G models meet the latest ATEX standards.

The RadEye G Ex Series of Personal Radiation Detectors comprises four versions of intrinsically safe handheld devices for gamma and dose rate measurements for operation in and around hazardous areas. 

  • Efficient and reliable dose and dose rate measurements.
  • Intrinsically safe according to ATEX standards.
  • Large, clear and backlit display for error free readings.
  • Rugged and reliable.
  • Lightweight, only 160g (5.6 oz.).
  • Low power technology.
Recommended for:
  • Emergency services
  • Fire brigades
  • Hazmat teams
  • Locations with risk of explosion
  • Refineries
  • Oil platforms

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

II 2G Ex ia IIB T4 IBExU10ATEX1096
For Cs-137 (622keV): 17 cps per mR/h
Energy-compensated Geiger-Mueller tube
45 keV to 1.3 MeV (+/-30%)
up to 1000 R
IP 65 according to EN 60529
R and R/h
0.41 lb.
2 AAA
RadEye G-Ex with Orange Label
900 hr.
50µR/h to 10R/h
1000 R/h
Personal Radiation Detector
3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
0.18 kg
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit