Thermo Scientific™ RadEye™ GR Grapple Monitoring System Available on GSA/VA Contract for Federal Government customers only.

Minimize the threat of radioactive material in the scrap metal stream with a wireless radiation detection system designed for grapple installation.

Manufacturer: Thermo Scientific™ 425522005

 View more versions of this product

Catalog No. 19-175-648

  Check Availability
Add to Cart

    Specifications

    Interface Board IR (standard) or Bluetooth (option)
    Audible Alarm 85 dB in 3 cm distance ( high power LED)
    Includes System Components: RadEye R Receiver Unit (option to increase the number of Receiver Units) ; Detector Unit (Installed on grapple) ; Battery Module and Radio Transceiver (Installed on grapple) ; Optional Datalogger
    Item Description Gooseneck adapter
    Communication Details ZigBee transmitter, 1 mW, >Range 100m, 328 ft.
    Data Management Details Internal Data Logger; RadEye PC-software compliant
    Description Gooseneck adapter

    In the metal recycling industry, discarded radioactive materials can seriously contaminate a shredder, baler, steel melt or bag house, resulting in significant clean-up costs and lost productivity. The Thermo Scientific™ RadEye™ GR Grapple Monitoring System is a wireless radiation detection system designed to be installed on a grapple to monitor scrap metal while it is still in the scrap pile under the grapple. Even when radiation is highly attenuated, the RadEye GR system responds to shielded radioactive sources to stop them from making their way into the steel making process.

    The RadEye GR Grapple Monitoring System is a radiation detection device designed and proven through extensive testing for the extreme forces and harsh conditions experienced when installed in a grapple. The RadEye GR system provides an additional, significant security increase to present portal monitoring because it is closer to the radioactive source and maintains contact for a longer period of time.

    • Compact detector size - negligible load capacity reduction while providing high sensitivity.

    • Enhanced sensitive for low gamma energies.

    • Superior value: Small investment and low cost of ownership.

    • Multiple portable RadEye GR display units possible.

    • Extremely high battery lifetime.

    • Nearly maintenance-free.

    • Very straightforward installation process.

    • Simple and comprehensive data logging and reporting.

    • Radiation test handled by single person.

    • ViewPoint centralized real time monitoring option.

    • Optimized shape and thickness of the detector dome allows even low energetic radiation to cause a significant signal in the detector.

    System Components:

    • RadEye R Receiver Unit (option to increase the number of Receiver Units).

    • Detector Unit (Installed on grapple).

    • Battery Module and Radio Transceiver (Installed on grapple).

    • Optional Datalogger.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit