Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RadHalo Spectroscopic Area Monitor Radio Communication Options Available on GSA/VA Contract for Federal Government customers only.

For use with RadHalo Spectroscopic Area Monitor models with the radio option

Manufacturer:  Thermo Scientific MTXR24L

 View more versions of this product

Catalog No. 19-175-825


  Check Availability
Add to cart

Specifications

Description

Matrix Base Radio and Matrix Radio Repeater
  • Include rugged IP67 housing, 2.4 GHz radio, external antenna connection, power connection, ethernet connection and wall mount
  • Require an appropriate 2.4 GHz antenna kit
  • Do not include a pole mounting kit or screws/anchors for wall mount
The Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit includes complete kit for mounting radios to a pole up to 12 inches in diameter.
Description & Specifications

Specifications

Matrix Radio Repeater - Dual 2.4 GHz Laird Radios
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit