Learn More
Andwin Scientific RADIACWASH 1GAL/21876302
SDP
Manufacturer:  Andwin Scientific 021876302
Encompass_Preferred
The Radiacwash™ Decontaminant provides secure elimination of radioactive contaminants from skin, cloth, metal, glass, floors, plastic, and equipments. This product can also get rid of pollutants like soil, grease, and residue. It is efficient and economical. It is packaged and sold in a 1 gal pour bottle.
Catalog No. NC9782031
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.