Learn More
Biodex Medical Systems RADIACWASH 1GAL
Manufacturer:  Biodex Medical Systems 005100
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Radiacwash - Gallon,1 Gallon,Radiacwash is a concentrated solution designed to rapidly control radioactive contamination and remove radioactive particles from surfaces by a two-way action, Radiacwash has a neutral pH, contains no phosphates
Catalog No. NC9633347
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »