Learn More
Andwin Scientific RADIACWASH DECONTAMINANT 1L
SDP
Manufacturer:  Andwin Scientific 21876347
Encompass_Preferred
The Radiacwash™ Decontaminant provides secure elimination of radioactive contaminants from skin, cloth, metal, glass, floors, plastic, and equipments. This product can also get rid of pollutants like soil, grease, and residue. It is efficient and economical. It is packaged and sold in a 1 L spray bottle.
Catalog No. NC9370849
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »