Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Radioactive Uncalibrated Solid Sources
Individual 1 in. uncalibrated solid source.
Supplier: Spectrum Techniques Inc TL204S1
Description
- Fabricated in 1 in. dia. (25mm) by 0.25 in. thick (6mm) plastic disk
- Disk sources are identified as alpha, beta or gamma
- Supplied individually or as a set
- Contains ±20% of the stated activity and is routinely compared to calibrated reference standards
Specifications
Radioactive Uncalibrated Solid Sources |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction