Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rae-1 epsilon Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene Symbol RAET1E
Name Recombinant Rae-1 epsilon Protein
Product Type Recombinant Protein
Source Baculovirus
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60207100U View Documents Novus Biologicals™
NBP260207100UG
100μg Each for $1,025.00
Add to cart
 
NB6020720UG View Documents Novus Biologicals™
NBP26020720UG
20μg Each for $287.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 31-225 of Human Rae-1 epsilon The Recombinant Human ULBP-4/RAET1E Protein is derived from Baculovirus. The Recombinant Human ULBP-4/RAET1E Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Recombinant Rae-1 epsilon Protein
Baculovirus
0.5mg/mL
SDS-Page
RAET1E
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit