Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rae-1 Rat anti-Mouse, DyLight 755, Clone: RM0511-14R4, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241749IR

Catalog No. NBP241749IR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Rae-1
RM0511-14R4
Flow Cytometry
Purified
Rat
IgG2
0.1 ml
Primary
135250
Mouse
Flow Cytometry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CD3E
Recombinant Mouse Rae-1delta; aa29-aa227.
Protein G purified
Store at 4C in the dark.
Monoclonal
This antibody recognizes mouse Rae-1 alpha, beta, gamma, delta, and epsilon by ELISAs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit