Learn More
Rae-1 Rat anti-Mouse, R-PE, Clone: 186107, R&D Systems
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB17582P100UG
Catalog No. FB17582P100
Add to cart
This item is not returnable. View return policy
Description
Description
Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Rae-1
186107
Flow Cytometry 0.25-1 μg/106 cells
Q9JI58
Raet1a
Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta , Leu29-Ser227, Accession No. Q9JI58
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse Rae-1. It recognizes Rae-1 alpha , beta , delta , gamma and epsilon .
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Conjugated, Purified, Supernatant
Rat
IgG2a
100 μg
Primary
8480
Mouse