Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAE1 gamma Rat anti-Mouse, PE, Clone: CX1, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 12588181

Catalog No. 50-112-2339


Add to cart

Description

Description

Description: The CX1 monoclonal antibody reacts with the mouse retinoic acid early inducible gamma (RAE-1gamma) and weakly cross-reacts with the RAE-1alpha and beta, but not with delta and ε. These 5 members of the RAE-1 family, along with another protein encoded by the minor histocompatibility gene complex called H60 have been identified as ligands for the mouse NKG2D molecule. These ligands are normally absent from adult tissue, however the presence of retinoic acid or certain disease conditions (such as tumors) can induce up-regulation of this protein at the cell surface. RAE-1alpha, -beta, and -gamma and H60 are expressed in BALB/c mice, but not in C57BL/6, whereas RAE-1delta, and -ε are expressed in C57BL/6 mice. It is reported that in NOD mice, both NKG2D and its ligand RAE-1 are expressed on the same NK cells, resulting in self-modulation of NKG2D expression and function. Applications Reported: The CX1 antibody has been reported for use in flow cytometric analysis. Applications Tested: The CX1 antibody has been tested by flow cytometric analysis of YAC-1 cell line and mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered. Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

RAE1 gamma
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O08604
RP4-800J21.2, MIG14, MRNP41, Mnrp41, dJ481F12.3, dJ800J21.1, homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer), mRNA export factor, mRNA export protein, mRNA-associated protein MRNP 41, mRNA-binding protein, 41-kD, migration-inducing gene 14, rae1 protein homolog
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
CX1
PE
Raet1c
Liquid
Raet1c
IgG2b, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19370
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.