Learn More
RAE1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 204911AP
Catalog No. 50-173-2918
Add to cart
Description
Description
Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
RAE1
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P78406
MIG14, Mnrp41, mRNA export factor, MRNP41, RAE1, Rae1 protein homolog
Rabbit
IgG
150 μL
-20°C
Primary
8480
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RAE1
Liquid
RAE1
RAE1 Fusion Protein Ag12102
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human