Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raf-1 Rat anti-Human, HRP, Clone: DPR/2/2A/3A, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275950H

Catalog No. NBP275950H


Add to cart

Description

Description

Raf-1 Monoclonal antibody specifically detects Raf-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Raf-1
DPR/2/2A/3A
Western Blot
Conjugated, Purified
RAF1
Synthetic peptide GQRDpSSYpYWEIEAS coupled to glutaraldehyde
Protein G purified
Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction
Primary
5894.0
Western Blot
HRP
PBS with No Preservative
cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1
Rat
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit