Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAF1 (cRAF) [Y340D] [Y341D] Recombinant Human Protein

RAF1 (cRAF) [Y340D] [Y341D] Recombinant Human Protein

$510.00 - $1740.00

Specifications

Accession Number NP_002871, Y340D, Y341D
Expression System Baculovirus
Gene Alias RAF1 (cRAF) Y340D Y341D
KinaseFamily RAF Kinase Family
Molecular Weight 66.4 kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3805
Encompass
View Documents
Invitrogen™
PV3805
10μg
Each for $510.00
Add to cart
 
PR7307A
Encompass
View Documents
Invitrogen™
PR7307A
100 µg
Each for $1,740.00
Add to cart
 
Description

Description

RAF1 (cRAF) is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated RAF1 can phosphorylate and activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylates and activates the serine⁄threonine specific protein kinases ERK1 and ERK2. The RAF1 Y340D Y341D mutant is constitutively active.
Specifications

Specifications

NP_002871, Y340D, Y341D
RAF1 (cRAF) Y340D Y341D
66.4 kDa
GST Tag
Baculovirus
RAF Kinase Family
Kinase Assay
SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit