Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAGE Rabbit anti-Human, Clone: JF0975, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267095
Description
RAGE Monoclonal antibody specifically detects RAGE in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RAGE | |
JF0975 | |
Unconjugated | |
Purified | |
EC 2.7.11.22, MOK, MOK Protein Kinase, RAGE-1, renal tumor antigen, Renal tumor antigen 1 | |
MOK | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Vision | |
Human,Mouse,Rat |
Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
TBS (pH7.4), 1%BSA, 40% Glycerol with 0.05% Sodium Azide | |
5891 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title