Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAGE, Rabbit anti-Mouse,Rat, DyLight 680, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP203951FR

Catalog No. NB004766


Add to cart

Description

Description

RAGE Polyclonal specifically detects RAGE in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RAGE
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11.22, MOK, MOK Protein Kinase, RAGE-1, renal tumor antigen, Renal tumor antigen 1
Rabbit
IgG
0.1 mL
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Vision
Primary
Note: This antibody was raised against an amino acid sequence that is 100% conserved in human, however western blot testing shows that it does not detected the expected ∽45 kD band in human lung lysate.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
MOK
A portion of amino acid 350-400 of human RAGE was used as the immunogen.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit