Learn More
Invitrogen™ RAGE (STK30) Mouse ELISA Kit
Sandwich ELISA Kit
$594.00 - $2930.00
Specifications
Accession Number | Q9WVS4 |
---|---|
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Protein Family | Kinase & Inhibitors |
Description
The Mouse RAGE (STK30) ELISA quantitates Ms RAGE in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms RAGE. Principle of the method The Mouse RAGE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.Specifications
Q9WVS4 | |
Biotin | |
Kinase & Inhibitors | |
Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30L, CD30, CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CSF, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL- | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
26448 | |
<12% | |
HRP | |
Other Research | |
RAGE1, Rage, Stk30 | |
1 hr. 20 min. |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Mouse | |
Colorimetric Microplate Reader | |
Mok | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.