Learn More
RAI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170841AP
Catalog No. 50-173-1375
Add to cart
Description
Description
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It is highly similar to its mouse counterpart and is expressed at high levels mainly in neuronal tissues. The protein encoded by this gene includes a polymorphic polyglutamine tract in the N-terminal domain. Expression of the mouse counterpart in neurons is induced by retinoic acid. This gene is associated with both the severity of the phenotype and the response to medication in schizophrenic patients.
Specifications
Specifications
RAI1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q61818, Q7Z5J4
KIAA1820, RAI1, retinoic acid induced 1, SMCR, SMS
Rabbit
IgG
150 μL
-20°C
Primary
10743, 19377, 303188
Western Blot
Unconjugated
RAI1
Liquid
Rai1
RAI1 Fusion Protein Ag9046
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat