Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAIDD/CRADD Mouse anti-Human, Alexa Fluor™ 594, Clone: 14G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242688AF594

Catalog No. NP242688A94


Add to cart

Description

Description

RAIDD/CRADD Monoclonal antibody specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

RAIDD/CRADD
14G8
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified, Conjugated
CRADD
Recombinant human RAIDD/CRADD (1-199aa) purified from E. coli
Protein A purified
Apoptosis, Cancer, Cell Biology, Tumor Suppressors
Primary
8738.0
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain
Mouse
IgG1a Kappa
100 μL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit