Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAIDD/CRADD Antibody (14G8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242688FR
Description
RAIDD/CRADD Monoclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
RAIDD/CRADD | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CRADD | |
Recombinant human RAIDD/CRADD (1-199aa) purified from E. coli | |
100 μL | |
Apoptosis, Cancer, Cell Biology, Tumor Suppressors | |
8738 | |
Store at 4C in the dark. | |
IgG1a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
14G8 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction