Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Rainbow Buffer Kit 4, 7, 10

Color coded technical buffer kit of ready to use pH buffer solution in predosing bottles that assure you get the exact dose of buffer needed for pH calibration. Singe use dosing reduces secondary contamination and exposure to environmental evaporation.

Manufacturer:  Sartorius™ PYB2502S

 View more versions of this product

Catalog No. 14-558-084


Add to cart

Description

Description

  • Color coded technical buffer kit
  • Reliable calibration needed for accuracy and precision
  • Multiple volume kits 250mL and 1L
  • Easy to dispence with predosing bottle reduce secondary contamination and exposure to environmental evaporation
Specifications

Specifications

pH Calibration
3 x 250mL
4, 7, 10
3 x 250mL Bottles
Self-Dosing Bottles
pH buffer
Undesignated
Documents
Promotions

Promotions

  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit