Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Rainbow™ Molecular Weight Markers: PROMO

Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining. GE Healthcare Life Sciences™ Rainbow™ Molecular Weight Markers use bright, distinctive colors to allow easier confirmation of transfer to blotting membranes and orientation.

Specifications

Color Multicolor
Format Liquid
For Use With (Application) Protein electrophoresis
Storage Requirements −15° to −30°C
Sufficient For 50 Minigels or 25 large gels
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Product Type Range Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Product Type Range Price Quantity & Availability  
45-001-592LR View Documents GE Healthcare
RPN756EPM
1.5 mg/mL Protein Molecular Weight Marker/Ladder 12000 to 225000Mr This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
45-001-593LR View Documents GE Healthcare
RPN755EPM
1 mg/mL Protein Molecular Weight Marker/Ladder 3500 to 38000Mr This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination.

  • High intensity, prestained multicolored bands allow distinction of closely related proteins
  • Full-Range Rainbow Markers (Mr 12,000 to 225,000) use ten separate proteins with six different colors
  • High-Range Rainbow Markers (Mr 12,000 to 225,000) use eight separate proteins with six different colors
  • Low-Range Rainbow Markers (Mr 3500 to 38,000) use seven separate proteins with five different colors
  • For use on SDS-PAGE

  • The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.
  • Markers are a mixture of individually colored proteins that are combined to produce bands of equal color intensity. Approximate protein concentration is 1 to 2 mg/mL, depending on the product
  • Each 250μL pack size is sufficient for use with 50 minigels (10 × 8cm) or 25 large gels (20 × 20cm)

Specifications

Specifications

Multicolor
Protein electrophoresis
50 Minigels or 25 large gels
Liquid
−15° to −30°C
Safety and Handling

Safety and Handling

ShelfLife : 3 Months

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit