Learn More
Andwin Scientific RAININ 10ML TIPS PK/200
SDP
Supplier:  Andwin Scientific RC10ML
Encompass_Preferred
The 10 mL tips manufactured by Rainin are confirmed to be BioClean; the tips are completely free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors, additives and bioactive contaminants. This product is packaged in resealable zip-lock bags that permit quick manual refills of racks. It is sold in a pack of 200.
Catalog No. NC0040649