Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

RALBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen RALBP1
Applications Immunohistochemistry, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093123
View Documents
Novus Biologicals
NBP303710100UL
100 μg Each for $320.71
N/A
NB093124
View Documents
Novus Biologicals
NBP30371020UL
20 μg Each for $138.37
N/A
Description

Description

RALBP1 Polyclonal antibody specifically detects RALBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, KnockDown
Specifications

Specifications

RALBP1
Unconjugated
Rabbit
Human, Mouse, Rat
Dinitrophenyl S-glutathione ATPase, ralA binding protein 1, ralA-binding protein 1,76 kDa Ral-interacting protein, RalBP1, Ral-interacting protein 1, RIP, RIP1, RLIP1, RLIP76DNP-SG ATPase
Recombinant fusion protein containing a sequence corresponding to amino acids 396-655 of human RALBP1 (NP_006779.1).
Polyclonal
Affinity purified
Immunohistochemistry, KnockDown, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
10928
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit