Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RALGDS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116790
Description
Positive Samples: U-87MG, Mouse ovary, Mouse lung Immunogen sequence: CNSSSALPLY NQQVGDCCII RVSLDVDNGN MYKSILVTSQ DKAPAVIRKA MDKHNLEEEE PEDYELLQIL SDDRKLKIPE NANVFYAMNS TANYDFVLKK RTFTKGVKVK HGASSTLPRM KQKGLKIAKG IF.
Stimulates the dissociation of GDP from the Ras-related RalA and RalB GTPases which allows GTP binding and activation of the GTPases. Interacts and acts as an effector molecule for R-Ras, H-Ras, K-Ras, and Rap.
Specifications
RALGDS | |
Polyclonal | |
Unconjugated | |
RALGDS | |
Gnds; KIAA1308; mKIAA1308; Ral guanine nucleotide dissociation stimulator; ral guanine nucleotide exchange factor; RALGDS; RalGEF; RGDS; RGF | |
Rabbit | |
Affinity chromatography | |
RUO | |
19730, 29622, 5900 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.04 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q03385, Q03386, Q12967 | |
RALGDS | |
Recombinant fusion protein containing a sequence corresponding to amino acids 771-902 of human RALGDS (NP_001258705). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction