Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RALGDS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549099

Catalog No. PA549099


Add to cart

Description

Description

Stimulates the dissociation of GDP from the Ras-related RalA and RalB GTPases which allows GTP binding and activation of the GTPases. Interacts and acts as an effector molecule for R-Ras, H-Ras, K-Ras, and Rap.
Specifications

Specifications

RALGDS
Polyclonal
PBS with 0.09% sodium azide
Q12967
Ral guanine nucleotide dissociation stimulator, RalGDS, Ral guanine nucleotide exchange factor, RalGEF, RALGDS, KIAA1308, RGF
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5900
Western Blot
Unconjugated
RALGDS
Liquid
RALGDS
KLH conjugated synthetic peptide between 531-559 amino acids from the Central region of human RALGDS.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit