Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RALGPS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144716
Description
Immunogen sequence: NMMCQLSVVE SKSATFPSEK ARHLLDDSVL ESRSPRRGLA LTSSSAVTNG LSLGSSESSE FSEEMSSGLE S Antigen sequence identity: Mouse 92%; Rat 90%.
RALGPS1 gene ontology annotations related to this gene include regulation of Ral protein signal transduction; small GTPase mediated signal transduction.
Specifications
RALGPS1 | |
Polyclonal | |
Unconjugated | |
RALGPS1 | |
5830418G11Rik; AI853783; AI854138; Kiaa0351; mKIAA0351; Ral GEF with PH domain and SH3 binding motif 1; ral GEF with PH domain and SH3-binding motif 1; Ral guanine nucleotide exchange factor 2; Ral guanine nucleotide exchange factor RalGPS1A; ralA exchange factor RalGPS1; RalGEF 2; RALGEF2; RALGPS1; Ralgps1a; ras-specific guanine nucleotide-releasing factor RalGPS1; si:dkey-191c17.1; zgc:165535 | |
Rabbit | |
Affinity chromatography | |
RUO | |
9649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JS13 | |
RALGPS1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 346-416. Recombinant protein control fragment (Product #RP-109724). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction