Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RALGPS2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | RALGPS2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP152930
|
Novus Biologicals
NBP152930 |
100 μL |
Each of 1 for $436.00
|
|
Description
RALGPS2 Polyclonal specifically detects RALGPS2 in Human samples. It is validated for Western Blot.Specifications
RALGPS2 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
Q86X27-2 | |
55103 | |
Synthetic peptides corresponding to RALGPS2(Ral GEF with PH domain and SH3 binding motif 2) The peptide sequence was selected from the C terminal of RALGPS2. Peptide sequence YLGIYLSDLTYIDSAYPSTGSILENEQRSNLMNNILRIISDLQQSCEYDI. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dJ595C2.1, FLJ10244, FLJ25604, KIAA0351, Ral GEF with PH domain and SH3 binding motif 2, Ral GEF with PH domain and SH3-binding motif 2, RalA exchange factor RalGPS2, Ral-A exchange factor RalGPS2, ras-specific guanine nucleotide-releasing factor RalGPS2 | |
RALGPS2 | |
IgG | |
Affinity Purified | |
32 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title