Learn More
Selleck Chemical LLC
Raloxifene HCl 100mg 82640-04-8 LY156758 (Keoxifene) HCl
Manufacturer: Selleck Chemical LLC S1227100MG
Raloxifene (LY156758, Keoxifene) HCl a selective and orally active estrogen receptor modulator (SERM), which inhibits human cytosolic aldehyde oxidase-catalyzed phthalazine oxidation activity with IC50 of 5.7 nM.