Learn More
Aviva Systems Biology Corp RALY Antibody - middle region (ARP33107_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP33107_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RALY. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RALY. Alternate names: HNRPCL2, MGC117312, P542ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62829