Learn More
RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Serum
No “scooping” needed, allowing for high-quality, tissue-free samples
Manufacturer:  RAM Scientific, Inc. 07 7450
 View more versions of this product
Catalog No. 14-915-61
Due to product restrictions, please to purchase or view availability for this product.
Includes: Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube
Specifications
Description
  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For plasma determinations in chemistry
  • Optional gel separator serves as a barrier between blood and serum after centrifugation
  • Available in a variety of sizes and draw volumes, with or without gel separator
  • Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Description & Specifications
Specifications
Yes
200μL
Standard Pierceable Cap
Clot Activator
Chemistry/Serum Tube
Intended for chemistry determinations that require serum or clotted blood samples. Amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin). Gel separator serves as a barrier between blood and serum after centrifugation.
Plastic Tube
Amber Microtube with Red Cap