Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson Gloves Ram Tough Welding Gloves

Catalog Number 17099886
Click to view available options
Size:
Medium
Large
X-Large

Ram Tough Welding Gloves have full-grain goatskin leather palm, split cowhide back, gunn cut design with straight thumb and leather reinforced pulse protector.

  • Full-grain goatskin leather palm
  • Split cowhide back
  • Gunn cut design with straight thumb
  • Leather reinforced pluse protector
  • Stitched with Aramid Fiber thread
  • Gauntlet style cuff with leather gore
  • Unlined
Back Material Split Cowhide Leather
Cuff Material Leather Gore
Color Gray
Grade Premium
Hand Ambidextrous
Material Goatskin Leather
Product Type MIG/TIG Welding Gloves
Size Medium
Style Gunn
For Use With (Application) Welding
Gender Men's
Glove Cuff Color Gray
Glove Cuff Style Gauntlet
Palm Type Full Grain Goatskin
Series Heat Wave
Thumb Style Straight
Length (English) 12 in.
Length (English) Cuff 5 in.
Length (Metric) 30 cm
Length (Metric) Cuff 13 cm
Show More Show Less
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit