Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ran Antibody (8D1A6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$560.50
Specifications
Antigen | Ran |
---|---|
Clone | 8D1A6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61832
![]() |
Novus Biologicals
NBP261832 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
Ran Monoclonal specifically detects Ran in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Ran | |
1 mg/mL | |
Monoclonal | |
Purified | |
Membrane Trafficking and Chaperones, Stem Cell Markers | |
Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4 | |
RAN | |
IgG1 | |
Protein G purified |
8D1A6 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Monkey | |
5901 | |
Purified recombinant fragment of human RAN (AA: 1-216) expressed in E. Coli. | |
Primary | |
24.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title