Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Economy Class Packs: Vertebrates
SCI_ED

Manufacturer:  Ranaco Corporation 150.2060.1

 View more versions of this product

Catalog No. S06930

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Economy Class Pack includes 30 specimens (usually in three ten packs) and a set of 15 dissection guides, all at one economical price.

  • Rat Pack: 7-9 in. plain preserved rats; An Illustrated Dissection Guide to the Rat (34 pgs.)
  • Frog Pack: 4- 5 in. plain grassfrogs; An Illustrated Dissection Guide to the Frog (16 pgs.)
  • Perch Pack: 7- 9 in. plain perch; An Illustrated Dissection Guide to the Mexican Gray Perch (24 pgs.)

Specifications

Specifications

Three ten packs of 7 to 9 in. plain preserved Rats, fifteen copies of 24 page and 12 figure guides set
Perch economy class pack
Each
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit