Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Economy Class Packs: Vertebrates
SCI_ED

$274.69

Specifications

Quantity Each
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
S06928
SCI_ED
View Documents
Ranaco Corporation
150.2040.1
Frog economy class pack Three ten packs of 4 to 5 in. plain Grassfrogs, fifteen copies of 16 page and 14 figure guides set
Each for $274.69
Add to cart
 
S06930
SCI_ED
View Documents
Ranaco Corporation
150.2060.1
Perch economy class pack Three ten packs of 7 to 9 in. plain preserved Rats, fifteen copies of 24 page and 12 figure guides set This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Economy Class Pack includes 30 specimens (usually in three ten packs) and a set of 15 dissection guides, all at one economical price.

  • Rat Pack: 7-9 in. plain preserved rats; An Illustrated Dissection Guide to the Rat (34 pgs.)
  • Frog Pack: 4- 5 in. plain grassfrogs; An Illustrated Dissection Guide to the Frog (16 pgs.)
  • Perch Pack: 7- 9 in. plain perch; An Illustrated Dissection Guide to the Mexican Gray Perch (24 pgs.)

Specifications

Specifications

Each
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit