Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP1 Monoclonal Antibody (OTI8E1)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527490

Catalog No. PIMA527490


Only null left
Add to Cart

Description

Description

RANBP1 Monoclonal Antibody for Western Blot

Ran/TC4-binding protein, RanBP1, interacts specifically with GTP-charged RAN. RANBP1 encodes a 23-kD protein that binds to RAN complexed with GTP but not GDP. RANBP1 does not activate GTPase activity of RAN but does markedly increase GTP hydrolysis by the RanGTPase-activating protein (RanGAP1). The RANBP1 cDNA encodes a 201-amino acid protein that is 92% similar to its mouse homolog. In both mammalian cells and in yeast, RANBP1 acts as a negative regulator of RCC1 by inhibiting RCC1-stimulated guanine nucleotide release from RAN.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RANBP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P43487
RANBP1
Full length human recombinant protein of RANBP1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI8E1
Unconjugated
RANBP1
HpaII tiny fragments locus 9A; hpaII tiny fragments locus 9a protein; Htf9a; Htf9-a; Htf9-a/RanBP1; RAN binding protein 1; ran-binding protein 1; RANBP1; RANG; Ran-specific GTPase-activating protein; testis secretory sperm-binding protein Li 221n
Mouse
Affinity chromatography
RUO
5902
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.