Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RanBP16 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | RanBP16 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111187
|
Invitrogen™
PA5111187 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111268
|
Invitrogen™
PA5111268 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: SQPPEKQQAM HLCFENLMEG IERNLLTK.
The transport of protein and large RNAs through the nuclear pore complexes is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN plays a key role in NLS-dependent protein import. RAN-binding protein-16 is a member of the importin-beta superfamily of nuclear transport receptors.Specifications
RanBP16 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIA9 | |
23039 | |
Recombinant Protein corresonding to Human RanBP16. Recombinant protein control fragment (Product #RP-100395). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Xpo7 | |
4930506C02Rik; BB164534; EXP7; exportin 7; exportin-7; Kiaa0745; mKIAA0745; RAN binding protein 16; ran-binding protein 16; Ranbp16; Xpo7 | |
Xpo7 | |
Primary | |
Antigen Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title