Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RanBP3 Antibody (12E11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242672AF647
Description
RanBP3 Monoclonal specifically detects RanBP3 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
RanBP3 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RANBP3 | |
Recombinant human RANBP3 (235-445aa) purified from E. coli | |
100 μL | |
Signal Transduction | |
8498 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
1200000000000 | |
Western Blot, ELISA | |
DKFZp586I1520, RAN binding protein 3, ran-binding protein 3, RAN-binding protein-3, RanBP3 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction